Would you like to log in again?
PIG® Traffic Mat® Absorbent Rug, New Pig
Supplier: New Pig
TRAFFIC MAT®
MAT218
56620-072EA 398.52 USD InStock
56620-072
PIG® Traffic Mat® Absorbent Rug, New Pig
Spill Control Materials
100% polypropylene material is heat fused and needle punched for incredible durability for walk-on and wheeled traffic
  • Absorbs and retains oils, coolants, solvents, and water
  • Dark gray color hides grime, so mat stays on the job longer
  • Rolls are ideal for walkways and large areas to soak up leaks and overspray
  • Cuts easily with scissors or utility knife
  • Can be incinerated after use to reduce waste or for fuels blending
Our most durable mat won't rip, shred, or fray, even under heavy forklift traffic
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty