Would you like to log in again?
PIG® HazMat Chemical Absorbent Pillow, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIL307 HR7015
56620-184CS 241.84 USD InStock
56620-184 56620-085
PIG® HazMat Chemical Absorbent Pillow, New Pig
Spill Control Materials
Absorbs the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide. Chemical-resistant pillow won't degrade or cause a dangerous reaction upon contact with corrosive spills. Pillow has large surface area, high capacity and fast-wicking filler to quickly soak up liquids; great for adding absorbency during spill response. Polypropylene skin resists chemicals and tearing; reduces dust and holds in liquid, even when saturated. Polypropylene filler is highly absorbent for containing corrosive or reactive spills.
  • Made of 100% chemical-resistant skin and fast-wicking filler
  • Large surface area and high capacity to quickly soak up liquids
Absorbs and retains most acids, bases and unknown liquids. Pink color is easily distinguishable to assure workers use the correct pillow during a spill emergency. Can be incinerated after use to reduce waste.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty