Would you like to log in again?
PIG® Absorbent Mat Roll, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MAT193 MAT202-01 MAT245 MAT246 MAT225 MAT153 MAT137 MAT220 MAT2009 MAT411
89205-892EA 389.59 USD InStock
89205-892 89073-832 56620-196 56620-042 56620-030 89073-830 89205-894 89073-818 56620-796 56620-038
PIG® Absorbent Mat Roll, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind.
  • Eight layers of 100% polypropylene thermally bonded to be exceptionally strong
  • Won't rip, tear or fray even when saturated
  • Dimple pattern speeds wicking of liquid for faster, easier cleanup
  • Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind
Absorbs and retains oils, coolants, solvents and water
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups. Mediumweight construction offers great performance for everyday tasks and cleanups. Light-weight construction is ideal for small tasks and cleanups.
Dark gray color hides grime so mat stays on the job longer. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Can be wrung out and incinerated after use to reduce waste or for fuels blending. Rolls are ideal for covering large areas and soaking up leaks and overspray. Easy-tear perforations let you take only what you need, so you use less mat and save money.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty