Would you like to log in again?
PIG® HazMat Spill Kit in Extra-Large Response Chest, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIT304-01 KIT304 KIT304-02
56620-462EA 3777.45 USD InStock
56620-462 56620-460 56620-216
PIG® HazMat Spill Kit in Extra-Large Response Chest, New Pig
Spill Control Materials
Seamless, polyethylene, lockable container keeps absorbents clean, dry and secure before use
  • Prepacked chest provides easy access to contents
  • Large, color-coded label makes identification fast and accurate
  • Tamper-proof seals help prevent pilfering of spill response supplies
  • Four-way forklift entry for easy moving
The 10" no flat wheels roll easily over uneven surfaces. The 5" rubber wheels are well suited to smooth concrete or pavement. Also available without wheels. Not UN rated.
PIG® HazMat Socks stop spreading spills; PIG® HazMat Pads and Pillows absorb; PIG® HazMat Pulp cleans up residue on rough surfaces. PIG® HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty