Would you like to log in again?
Accessories for PIG® HazMat Spill Kit in 95-Gallon Overpack Salvage Drum, New Pig
Supplier: New Pig
KITR302
56620-284EA 1393.32 USD InStock
56620-284
Accessories for PIG® HazMat Spill Kit in 95-Gallon Overpack Salvage Drum, New Pig
Spill Control Materials
Refill for PIG® HazMat Spill Kit in 95-Gallon Container. Don't be caught with an empty spill kit when you need it most! Keep a refill handy to instantly replace used contents.
  • Packed to refill your PIG HazMat Spill Kit in 95-Gallon Overpack Salvage Drum or 95-Gallon Container
  • PIG HazMat Socks stop spreading spills; PIG HazMat Pads and Pillows absorb quickly
  • PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Temporary disposal bags help make cleanup easier
  • Tamperproof seals help prevent pilfering of spill response supplies
Only PIG Spill Kit Refills feature PIG Absorbents proven for rapid response.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty