Would you like to log in again?
PIG® Oil-Only Spill Kit in 95-Gallon Wheeled Overpack Salvage Drum, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIT441-WH
56620-366EA 1028.76 USD InStock
56620-366
PIG® Oil-Only Spill Kit in 95-Gallon Wheeled Overpack Salvage Drum, New Pig
Spill Control Materials
Overpack drum container is UN Rated for shipping waste after oil spill cleanup
  • Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole
  • Large, color-coded label makes identification fast and accurate
  • Ledges molded into container make kit easy to move by forklift
  • Tamperproof seals help prevent pilfering of spill response supplies
  • UN1H2/X113/S
Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail. Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry.
The 6" olefin wheels make spill kit highly mobile for speedy response anywhere in your facility. Lift-out, prepacked baskets speed access and guard contents from UV. PIG Oil-Only Booms stop spreading spills; PIG Stat-Mat absorbs quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty