Would you like to log in again?
PIG® Oil-Only Spill Kit in Extra-Large Response Chest, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIT404-01 KIT404-02 KIT404-03
56620-474EA 2429.36 USD InStock
56620-474 56620-368 56620-476
PIG® Oil-Only Spill Kit in Extra-Large Response Chest, New Pig
Spill Control Materials
Seamless, polyethylene, lockable container keeps absorbents clean, dry and secure before use
  • Prepacked, compartmentalized container provides easy access to contents
  • Large, color-coded label makes identification fast and accurate
  • Temporary disposal bags help make cleanup easier
  • Tamperproof seals help prevent pilfering of spill response supplies
  • Four-way forklift entry for easy moving
The 10" no flat wheels roll easily over uneven surfaces. The 5" rubber wheels are well suited to smooth concrete or pavement. Also available without wheels. Not UN rated.
PIG Oil-Only Booms stop spreading spills; PIG Stat-Mat Pads and Pillows absorb quickly. Absorbents meet NFPA 99 standards for static decay to allow use with fuels and flammables.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty