Would you like to log in again?
PIG® Dark Oil-Only Absorbent Boom, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BOM415 BOM414 BOM413 BOM410 BOM412 BOM411
56620-380PK 146.35 USD InStock
56620-380 56620-404 56620-424 56620-400 56620-422 56620-378
PIG® Dark Oil-Only Absorbent Boom, New Pig
Spill Control Materials
Dark color makes booms less noticeable when deployed on land or water
  • Absorbs and retains oils and oil-based liquids, including lubricants and fuels, without taking in a drop of water
  • Strong, zinc-plated carbon steel attachment clips and rings allow you to link booms together for greater length
  • Can be incinerated after use to reduce waste or for fuels blending
Floats to confine and soak up spills on water; hugs ground for land-based spills. Spunbond polypropylene skin is UV resistant up to 4 months. Tough outer mesh is UV resistant for long-term outdoor use without degradation; lets fluids easily pass through to filler material. Rope running along length of boom withstands up to 400-lb. loads, adding strength for deployment and retrieval. Booms float at surface for easy retrieval, even when saturated
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty