Would you like to log in again?
PIG® HazMat Spill Kit in 65-Gallon High-Visibility Container, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIT363
56620-446EA 926.19 USD InStock
56620-446
PIG® HazMat Spill Kit in 65-Gallon High-Visibility Container, New Pig
Spill Control Materials
Lift-out, prepacked baskets speed access and guard contents from UV. PIG HazMat Socks and Dikes stop spreading spills; PIG HazMat Pads and Pillows absorb quickly. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide.
  • Lift-out, prepacked baskets speed access and guard contents from UV
  • Socks and Dikes stop spreading spills
  • Pads and Pillows absorb quickly
  • Absorbents are specially treated for concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
  • Not UN rated
Lime green drum is easy to locate
No wheels. Not UN rated.
Lightweight, 100% polyethylene drum resists chemicals and keeps contents clean and dry. Large, color-coded label makes identification fast and accurate. Easy-access threaded lid (not rated for shipping). Tamperproof seal helps prevent pilfering of spill response supplies.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty