Would you like to log in again?
PIG® Poly Modular Spill Deck, New Pig
Supplier: New Pig
PAK529 PAK531 PAK524
56620-614EA 1005.74 USD InStock
56620-614 56620-612 56620-610
PIG® Poly Modular Spill Deck, New Pig
Drum Storage
Unique "flow-through" sump design lets liquid pass freely from one sump to another to help you comply with containment regulations. Modular design features exclusive bulkhead fittings and predrilled ports to connect to other decks; ready to use as soon as they arrive. Mix and match the sections you need to fit your space and help meet sump capacity requirements. Special tools and hardware to connect sections is included with each system.
  • Unique "flow-through" sump lets liquid pass freely from one sump to another to comply with containment regulations
  • Bulkhead fittings and predrilled ports connect to other decks
  • Mix and match sections to fit your space and help meet sump capacity requirements
  • Special tools and hardware to connect sections included with each system
  • Low-density polyethylene (LDPE) resists UV rays, rust, corrosion and most chemicals
  • Textured grating adds traction and lifts out for easy sump access
  • Translucent white sidewalls allow easy visual inspection
Low-profile 5.75"H design is easy to load or unload; pumps and funnels are easy to access
Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals. Textured grating adds traction and lifts out for easy sump access. Translucent white sidewalls allow easy visual inspection.
Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty