Would you like to log in again?
PIG® Steel Loading Ramp, New Pig
Supplier: New Pig
PAK538
56620-616EA 494.99 USD InStock
56620-616
PIG® Steel Loading Ramp, New Pig
Drum Storage
Heavy-duty steel ramps are built strong for big loads. Skip the forklift and roll your drums up a ramp.
  • Heavy-duty steel construction is strong and durable
  • Adjustable legs for use with a range of pallet and deck heights
  • Edge guards guide drum trucks and help prevent them from rolling off the sides
  • Lip of ramp secures to edge of pallet or deck
Note: Ramp must be detached to close doors. Diamond plate surface provides traction for added safety.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty