Would you like to log in again?
PIG® Conical Drain Plug, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PLR208 PLR210 PLR209 PLR252
56620-658EA 245.68 USD InStock
56620-658 56620-656 56620-654 56620-652
PIG® Conical Drain Plug, New Pig
Spill Control Materials
Durable polyurethane plug forms a liquid-tight, chemical-resistant seal in seconds.
Just wedge plug in drain for an easy alternative to permanently sealing your drains with cement. Installed plug is low-profile enough to fit below floor grate. Allows your drain to function normally when plug is not in place; removes easily with hand-tools.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty