Would you like to log in again?
PIG® Barrel Top Absorbent Mat, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG®
MAT244 MAT208 MAT237 MAT255
89073-834CS 68.85 USD InStock
89073-834 56620-678 89073-836 56620-036
PIG® Barrel Top Absorbent Mat, New Pig
Spill Control Materials
Soak up pump drips without cluttering up your work space with dirty rags
  • Absorbs and retains oils, coolants, solvents, and water
  • Dark gray color hides grime, so mat stays on the job longer
  • Fits 55 gallon steel drum tops; pre-cut with holes for 3/4" and 2" bungs to provide a perfect fit
  • Dispenser box makes it easy to carry and store mats while protecting them from dirt and moisture
  • Heavy weight construction is highly durable and absorbent to handle pump drips and dispensing spills
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty