Would you like to log in again?
PIG® STAT-MAT® Absorbent Mat Roll, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG® Stat-Mat®
MAT402 MAT418 MAT420
56620-682EA 382.49 USD InStock
56620-682 56620-684 89073-838
PIG® STAT-MAT® Absorbent Mat Roll, New Pig
Spill Control Materials
STAT-MAT Pads and Rolls pass NFPA 99 standards for static decay and surface resistivity, making them perfect for cleaning up solvents and fuels
Packaged in special static-dissipative bags to help reduce static charge buildup. Floats to clean up spills on water. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Rolls are ideal for covering large areas and soaking up leaks and overspray. Easy-tear perforations let you take only what you need, so you use less mat and save money.
Heavyweight construction is highly durable and absorbent for demanding tasks and large-volume cleanups. Light-weight construction is economical for small tasks and cleanups.
Absorbs and retains oils and oil-based liquids - including acetone, benzene, gasoline, toluene, xylene, mineral spirits and jet fuels - without taking in a drop of water. Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated. Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup. Highly absorbent, fine-fiber construction won't leave behind liquids or fiber residue
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty