Would you like to log in again?
PIG® Absorbent Mat Pads, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MAT203 MAT216 MAT412 MAT231
56620-044PK 180.58 USD InStock
56620-044 56620-032 56620-726 56620-544
PIG® Absorbent Mat Pads, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to make PIG® Absorbent Mat Pads exceptionally strong; they won't rip, tear or fray even when saturated.
  • Polypropylene (made with 50% recycled content) soaks up and retains liquids
  • Dimpled for quicker absorption of water-based liquids, petroleum products, solvents, and broad range of chemicals
  • Choose light, medium, or heavyweight depending on the amount of liquid you need to absorb
  • Can be incinerated after use to reduce waste
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
Absorbs and retains oils, coolants, solvents and water. Dark gray color hides grime so mat stays on the job longer. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Can be wrung out and incinerated after use to reduce waste or for fuels blending. PIG® pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need so you use less mat and save money.
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups. Mediumweight construction offers great performance for everyday tasks and cleanups.
Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty