Would you like to log in again?
PIG® Latching Drum Lid for Open-Head Steel Drums, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG®
DRM659-BK DRM659-RD DRM821-RD DRM659-YW DRM821-YW DRM821-BK
56620-738EA 917.39 USD InStock
56620-738 89032-424 89032-428 89032-426 56620-742 56620-740
PIG® Latching Drum Lid for Open-Head Steel Drums, New Pig
Lids Lids for Laboratory Vessels/Containers
Latching handle easily opens and closes with one hand.
  • Nitrile gasket and locking mechanism seal lid, reduces vapor emissions keeps drum contents dry and secure
  • Easy-access lid closes securely to help you meet haz-waste and closed-container regs
Hinge stops keep funnel lid upright when open. Durable, chemical-resistant powder coat helps ensure long service life. Lockable latch helps prevent unauthorized access. Hinge pin is made for easy lid removal without loosening band around drum. Hinged with one hand latch.
Accessories information: Replaces gasket on model DRM659 (PIG Latching Drum Lid). Includes gasket and adhesive.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty