Would you like to log in again?
Respirator Filter Cartridges with Stacked P100 Filter 6000 Series, 3M™
Supplier: 3M
7000126166 7000126165 7000126166 7000051992 7000029640 7000002041 7000051992 7000002009 7000029639 7000029639 7000002041 7000126165 7000029640
56621-896CS 1869.33 USD InStock
CA56621-048 CA56621-894 56621-896 CA56621-896 56621-898 56621-892 CA56621-892 56621-048 56621-894 56621-904 CA56621-904 CA56621-898 67100-995
Respirator Filter Cartridges with Stacked P100 Filter 6000 Series, 3M™
Respirators Reusable Respirators Respirator Filters
For use with 3M™ 6000 series respirator facepieces.
  • NIOSH-approved P100 filter is permanently attached
  • Manufacturer’s time use restrictions of 40 hours of use or 30 days
  • Certain Organic Vapors/Acid Gas/P100 approved for use against chlorine, hydrogen chloride, sulfur dioxide, hydrogen fluoride, hydrogen sulfide (escape only)
  • Multi-Gas/Vapor/P100 approved for use against certain organic vapors, chlorine, chlorine dioxide, hydrogen chloride, ammonia, methylamine, sulfur dioxide, hydrogen fluoride, hydrogen sulfide (escape only)
NIOSH approved P100 filter is permanently attached. Manufacturer’s time use restrictions of 40 hours of use or 30 days, whichever is first, apply if used in environments containing only oil aerosols.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. OSHA regulations require that gas-proof goggles be worn with half facepiece respirators when used against formaldehyde.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty