Would you like to log in again?
8212 Disposable N95 Particulate Respirators, 3M™
Supplier: 3M
7000002027 7000002027
56621-926CS 434.69 USD InStock
56621-926 CA56621-926
8212 Disposable N95 Particulate Respirators, 3M™
Respirators Disposable Respirators Particulate Respirators
Designed for use when welding, torch cutting, soldering, brazing and working with molten metals
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Helps provide protection when welding on galvanized and stainless steel. Fits easily under a welding hood. The respirator filter is at least 95% efficient at removing solid and liquid particles (excluding those containing oil). Adjustable straps, face seal, and Cool Flow® exhalation valve provide added comfort and durability. Caking-resistant filter media construction allows for easier breathing over long periods of wear.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty