Would you like to log in again?
AIR HOSE HIGH PRESSURE PK1
Supplier: 3M OCCUPATIONAL HEALTH
High Pressure Air Hose^ Can be used on 3M* High Pressure Compressed Air Systems. It has a 3/8 inch inner diameter and is equipped with MPT on both ends to accept 1/4 inch FPT fittings. Coiled hoses should be used in single length only. They cannot be combined.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
AIR HOSE HIGH PRESSURE PK1 56621-990 7000031122 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty