Would you like to log in again?
VWR® SoftCIDE® Extra-Mild Hand Soap
Supplier: VWR
VWR® SoftCIDE®
21032-06-601640 21128-04-601640 21016-06-601640
56614-420CS 275 USD InStock
56614-420 56614-424 56700-124
VWR® SoftCIDE® Extra-Mild Hand Soap
Skincare Soaps and Skin Cleansers
Unique formulation helps replenish skin’s natural oils that are rinsed away by frequent washing.
  • Anti-microbial strength
  • Effective against broad spectrum of microorganisms
  • pH balanced for skin
Soap has antimicrobial strength but is extra mild on hands. Contains 1% chloroxylenol (PCMX), a fast-acting, broad spectrum antimicrobial ingredient that is effective against gram-negative and gram-positive bacteria as well as yeast.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty