Would you like to log in again?
PROBE OPEN-HNDL GRAIN 6-OPENNG
Supplier: SEEDBURO EQUIPMENT CO
Open Handled Probe^ Without partitions. Used by grain elevator operators to obtain samples from truck loads. Probe has 6 openings. Length: 101.6cm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PROBE OPEN-HNDL GRAIN 6-OPENNG 56724-005 20-OH Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty