Would you like to log in again?
PROBE AUGER TIP PARTTND 11OPNG
Supplier: SEEDBURO EQUIPMENT CO
Auger Tip Partitioned Probe^ Brass partitioned probe with wooden handle with a 30.5cm steel T-handle below. With 11 openings and an auger point to aid sampling wet or closely packed material. Shipping dimensions: 165x20.3x10.2cm.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PROBE AUGER TIP PARTTND 11OPNG 56724-045 552 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty