Would you like to log in again?
Zipper Seal Bags, LDPE, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
628415 628405 628345 628355 628435 628325 628365 628425 628335
30620-118CS 250.47 USD InStock
30620-118 56766-304 30620-116 56766-306 56766-300 56766-314 56766-302 30620-122 30620-120
Zipper Seal Bags, LDPE, Dynalon
Bags Sample Bags
LDPE zipper seal bags are ideal for collecting, storing, and protecting laboratory specimens and samples.
  • No printing on the bag allows for easier labeling and marking
  • Bags lock easily and securely with the zipper locking feature
Not suggested for use with liquids, bags provide many laboratory and industrial applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty