Would you like to log in again?
Coliwasa Tubes, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
76100-200
56925-225CS 564.02 USD InStock
56925-225
Coliwasa Tubes, Kimble Chase, DWK Life Sciences
Samplers Fluid Samplers
The Coliwasa tube is used to obtain a vertical column of liquid representing an accurate cross-section of the sampled material
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Two-part borosilicate glass tube has a ball in the lower end which acts as a check valve to prevent sample loss during transfer. Both parts are prescored, allowing the unit to be easily snapped apart for safe disposal.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty