Would you like to log in again?
Drum Samplers, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
76000-150 76000-25 76000-75
56925-234CS 385.45 USD InStock
56925-234 56925-230 56925-232
Drum Samplers, Kimble Chase, DWK Life Sciences
Samplers Fluid Samplers
Conventional open-end drum thieves provide a quick method for sampling unmarked drums
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Units are easily broken for quick disposal.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty