Would you like to log in again?
Scintillation Vial, Glass, without Screw Cap, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
986532
56941-604CS 310.5 USD InStock
56941-604
Scintillation Vial, Glass, without Screw Cap, Wheaton, DWK Life Sciences
Vials
Manufactured from low-potassium borosilicate glass to critical manufacturing tolerances, ensuring reliable, consistent results in scintillation counting.
  • Background counts remain low and ultraviolet transmission high
  • Tight-fitting caps have tops suitable for writing
  • Vials with attached or unattached screw-caps
Screw caps can be purchased separately.
Vials are designed to load properly into scintillation counting equipment without jamming or breaking.
Ordering information: Packaged in 5 utility trays with each partitioned tray holding 100 vials. Vials conform to ASTM E 438 Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty