Would you like to log in again?
AIR HOOD TYVEK DISPOSBLE LP
Supplier: ALLEGRO INDUSTRIES
Fully Disposable Tyvek* Hood with Low Pressure Flow Adaptation^ One-piece construction. Breathing tube and head suspension integrated with hood assembly. Fully disposable, eliminating need for decontamination. Flexible and splash resistant. Ideal for workers with beards or goggles.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
AIR HOOD TYVEK DISPOSBLE LP 57006-054 9910D Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty