Would you like to log in again?
BLENDER BASE F/BLENDER W/JAR
Supplier: EBERBACH CORP
Blender Base for Waring* Blenders^ Blender Base for Waring* Blenders, 70mm base for wide-mouth or mason jars. Comes with one borosilicate Glass straight-side wide mouth 250mL jar and small size blending assembly.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BLENDER BASE F/BLENDER W/JAR 57007-338 E8495.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty