Would you like to log in again?
BLADE ASSEMBLY STANDARD
Supplier: EBERBACH CORP
Waring* Blending Assembly^ 6cm stainless steel blades. For use with highly corrosive sample materials. Supplied with blades, bearing holder, shaft, bearing cap, and cap nut. Bearing is pure carbon. Cap, bearing seal washer, and blending assembly seal washer are of fluoropolymer resin.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BLADE ASSEMBLY STANDARD 57007-356 E8670.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty