Would you like to log in again?
Barnstead/Lab-Line Universal Platforms for Shakers, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Barnstead/Lab-Line
30120BI 30125 30128 30115BI 30124 30106 30100 30110 30105
72890-048EA 2116.12 USD InStock
72890-048 57018-990 13306-568 57019-031 57019-020 57019-304 97040-486 57018-913 97040-484
Barnstead/Lab-Line Universal Platforms for Shakers, Thermo Scientific
Shakers and Mixers Shaking Incubators
Platforms may be customized for specific shaking requirements
Platforms are drilled and tapped with the maximum number of holes for use with a variety of different types of vessels.
Ordering information: Platforms only. For clamps, see the Accessories for MaxQ™ 8000 Stackable Orbital Shakers, Thermo Scientific series. For tube racks for use with universal platforms, see the Barnstead/Lab-Line Unwire™ Test Tube/Microcentrifuge Tube Racks for Universal Platforms, Thermo Scientific series.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty