Would you like to log in again?
Barnstead/Lab-Line Clamps for Universal Platforms, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Barnstead/Lab-Line
30153 30155 30157BI 30152BI 30161 30162 30159 30150BI 30175 30160 30156BI 30151 30158 30154BI
57019-678EA 61.48 USD InStock
57019-678 57019-674 57019-696 57019-686 57019-676 490014-162 57019-688 57019-692 57019-670 57019-682 57019-672 57019-684 57019-690 57019-680
Barnstead/Lab-Line Clamps for Universal Platforms, Thermo Scientific
Shakers and Mixers Shaking Incubators
Use with Thermo Scientific universal platforms (57018-990 series)
Clamp 57019-692 accommodates a 2.5 l low-form culture flask. Clamp 57019-696 accommodates a 2.8 l Fernbach flask. Clamp 47742-748 accommodates a microplate or deep well plate. All other clamps are for Erlenmeyer flasks.
Clamps are constructed of stainless steel with retaining springs to hold vessels securely in place.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty