Would you like to log in again?
Holders for Nutating Shakers, Separatory Funnel, Glas-Col
Supplier: Glas-Col
099AVH0500S 099AVH1000S 099AVH2000S
57020-292EA 920.32 USD InStock
57020-292 57020-293 57020-291
Holders for Nutating Shakers, Separatory Funnel, Glas-Col
Support Stands
Specially designed for our floor model and mid range shakers.
  • Quick release front clamp for ease in filling of funnel
  • Design eliminates excessive handling of glassware
  • Mount to shaker platform with no fasteners
These separatory funnel holders have a rear support which remains stationary while front part holding the funnel can be released and rotated 360 degrees. This permits the funnel to be filled, inverted for shaking, and then reinverted for addition and/or extraction. All while the flask remains in the holder.
Ordering information: One holder holds one funnel.
Delivery information: 7 to 10 days after order.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty