Would you like to log in again?
Holder for Bench Top Reciprocating Shaker, Mojonnier Fat Extraction Flask, Glas-Col
Supplier: Glas-Col
099AMFG3
57020-299EA 821.5 USD InStock
57020-299
Holder for Bench Top Reciprocating Shaker, Mojonnier Fat Extraction Flask, Glas-Col
Supports Flask Supports
This flask holder lets you use our bench top shaker for Mojonnier fat extraction.
  • Two mojonnier flask holders can be shaken at once
  • Load maximum of up to 5 pounds need not be balanced
  • Use for vertical and horizontal shaking
Each holder accepts up to four flasks 37 mm in diameter. Unstoppered flasks may be shaken vertically. For horizontal shaking the flasks should be stoppered and held in with the built in retrainers.
Ordering information: One holder holds four flasks.
Delivery information: 7-10 days after order.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty