Would you like to log in again?
Recombinant Human IFN-gamma ELISA Standard, BioLegend
Supplier: BioLegend
570209 570209
570209-BLEA 108.51 USD InStock
570209-BL CA570209-BL
Recombinant Human IFN-gamma ELISA Standard, BioLegend
Assays ELISAs
This IFN-γ protein is useful as a standard for a human IFN-γ sandwich ELISA, using unlabeled NIB42 antibody or unlabeled MD-1 antibody for capture and biotinylated 4S.B3 antibody for detection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty