Would you like to log in again?
Light Task™ Supported Polyurethane-Coated Gloves, Honeywell Safety
Supplier: Honeywell Safety
Light Task™
NF15/9L NF15/9L NF15/11XXL NF15/8M-H5 NF15/10XL NF15/11XXL NF15/7S NF15/7S NF15/10XL
57100-329CS 827.28 USD InStock
CA57100-339 57100-329 CA57100-327 CA57100-337 57100-339 57100-327 CA57100-335 57100-337 57100-335
Light Task™ Supported Polyurethane-Coated Gloves, Honeywell Safety
Gloves
Ideal for detailed assembly, inspection, light fabrication, small parts handling, or tasks requiring maximum dexterity.
  • Polyurethane coating for excellent grip and fingertip sensitivity, nylon liner shell for minimal shrinkage when laundered
  • Uncoated area on back provides ventilation and comfort, knit wrist prevents material from entering glove
Gloves feature a polyurethane coating for excellent grip and fingertip sensitivity, and a nylon liner shell that displays minimal shrinkage when laundered. An uncoated area on the back of the glove provides ventilation and comfort, and the knit wrist prevents material from entering the glove.
Color: white
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty