Would you like to log in again?
3" Test Sieves, W.S. Tyler™
Supplier: W.S. Tyler
1539 1545 1303 1292 1298 1541 1557 1548 1296 1559 1549 1554 1294 1307 1300 1310 1547 1301 1553 1290 1538 1551 1304 1543 1299 1293 1540 1555 1306 1302 1295 1297 1542 1546 1305 1308 1544
89423-898EA 67.89 USD InStock
89423-898 30624-148 30624-146 30624-168 30624-176 30624-154 30624-174 30624-152 57321-950 30624-172 30624-150 30624-170 89424-002 89424-000 89424-006 89424-004 89424-008 30620-686 80700-020 80700-022 30624-138 30624-136 30624-158 30624-156 30624-178 30624-134 30624-144 30624-166 30621-290 30624-142 30624-164 30624-140 30624-162 89424-010 30624-160 89424-012 57321-945
3" Test Sieves, W.S. Tyler™
Sieves Test Sieves
These test sieves are available with stainless or brass frame and stainless cloth.
  • Depth to cloth: 1-1/4"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty