Would you like to log in again?
SIEVE US #325 FH BR SS 6
Supplier: W.S. Tyler
Test Sieve^ #325. Diameter: 15.2cm. Full height: 6.7cm. Depth to cloth: 4.4cm. Stainless steel construction.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SIEVE US #325 FH BR SS 6 57321-965 3884 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty