Full-height sieves are 8.3cm (31/4") high above stainless steel wire cloth
Each sieve is serialized and supplied with a “Test Sieve Certificate.” Sieves are marked with the USA Sieve Number. Sieves one millimeter and coarser are identified by opening in millimeters and inches; sieves finer, by opening in microns and inches.
Overall height: 10.8cm (41/4"). Half-height sieves are 4.0cm (15/8") high above stainless steel wire cloth. Overall height: 6.6cm (25/8"). Diameter: 30.5cm (12").
Certifications: Conform to ASTM-E11 specifications.