Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3" Sonic Sifter Sieves, Advantech

Supplier: Advantec
L3-S400 L3-S4 L3-S25 L3-S14 L3-S60 L3-S3.5 L3-S70 L3-S8 L3-S325 L3-S10 L3-S40 L3-S35 L3-S12 L3-S170 L3-S16 L3-S6 L3-S270 L3-S80 L3-S5 L3-S50 L3-S120 L3-S230 L3-S635 L3-S30 L3-S100 L3-S450 L3-S18 L3-S140 L3-S20 L3-S45 L3-S200 L3-S7 L3-S500
57333-760EA 177.02 USD InStock
57333-760 57333-782 57333-762 57333-780 57333-768 57333-785 57333-764 57333-766 57334-030 57334-012 57334-010 57334-016 57334-014 57334-018 57333-770 57333-772 57333-778 57333-756 57333-758 57333-774 57333-752 57333-754 57333-776 57334-020 57334-002 57334-024 57334-022 57334-000 57334-028 57334-006 57334-004 57334-026 57334-008
3" Sonic Sifter Sieves, Advantech
Sieves Test Sieves
These 3" diameter "see-through" acrylic framed sieves are made of stainless steel wire cloth and specifically designed to be used with the Sonic Sifter Separator.

  • Ideal for particles ranging from 20 µm to 5.6 mm

Centerline® Sieve Inspection or Calibration Certification reports provide detailed statistical data that is linked to each serialized sieve and becomes an integral part of your Quality Assurance Program.

Certifications: Conforms to ASTM E11 specification

Ordering information: Certifications are available as accessories.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty