Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Test Sieves 8" , Brass
Supplier: VWR
VWR®
170BB8H 30BB8F 60BB8F 8BB8H 16BB8H 14BB8F 45BB8H 20BB8H 20BB8F 18BB8F 35BB8F 270BB8H 50BB8F 35BB8H 14BB8H 10BB8H 325BB8H 100BB8F 40BB8H 16BB8F 70BB8F 120BB8H 12BB8F 50BB8H 270BB8F 200BB8F 18BB8H 30BB8H 25BB8F 40BB8F 100BB8H 70BB8H 60BB8H 140BB8H 230BB8H 25BB8H 120BB8F 230BB8F 8BB8F 80BB8F 12BB8H 325BB8F 200BB8H 170BB8F 140BB8F 45BB8F 10BB8F 80BB8H
57334-170EA 219.55 USD InStock
57334-170 57334-192 57334-190 57334-152 57334-196 57334-174 57334-130 57334-150 57334-172 57334-194 57334-112 57334-156 57334-178 57334-134 57334-110 57334-132 57334-176 57334-154 57334-116 57334-138 57334-158 57334-114 57334-136 57334-118 57334-180 57334-162 57334-184 57334-140 57334-160 57334-182 57334-188 57334-144 57334-166 57334-100 57334-122 57334-120 57334-186 57334-164 57334-142 57334-126 57334-104 57334-124 57334-168 57334-146 57334-102 57334-108 57334-106 57334-128
VWR® Test Sieves 8" , Brass
Sieves Test Sieves
VWR® 8" diameter (203.2 mm ) all brass test sieves are durable and ideal for sieving applications where a medium sample size is required.
  • Highly durable construction
  • Brass fabrication
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.
Accessories information: Half height (1") or full height (2") pans are available in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Ordering information: Sieves are available in half height (1"), or full height (2"). Sieve height is measured from the frame rim to the sieve cloth.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty