Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® 8" Test Sieves, Brass and Stainless Steel

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
1-1/4"BS8H 80BS8H 7BS8H 5BS8H 3/4"BS8H 4.24"BS8F 1-3/4"BS8F 100BS8F 140BS8F 5/16"BS8F 270BS8H 635BS8F 500BS8H 3/8"BS8H 1.00"BS8H 35BS8F 60BS8F 325BS8H 4"BS8H 1/2"BS8H 3.5BS8F 70BS8F 3-1/2"BS8F 8BS8F 2"BS8F 8BS8H 170BS8H 1-1/4"BS8F 3/8"BS8F 230BS8H 70BS8H 1-1/2"BS8H .265"BS8H 500BS8F 5/8"BS8F 25BS8F 200BS8H 16BS8H 7BS8F 35BS8H 12BS8H 1/4"BS8F 3/4"BS8F 7/8"BS8F 1/8"BS8F 5"BS8H .265"BS8F 10BS8H 50BS8H 100BS8H 18BS8F 50BS8F 400BS8F 1/2"BS8F 325BS8F 450BS8F 45BS8F 60BS8H 25BS8H 5"BS8F 4"BS8F 80BS8F 16BS8F 2.12"BS8F 2-1/2"BS8F 230BS8F 4BS8H 3.5BS8H 1-1/2"BS8F 5/16"BS8H 120BS8H 120BS8F 635BS8H 30BS8F 20BS8H 5/8"BS8H 30BS8H 140BS8H 10BS8F 40BS8H 4BS8F .530"BS8F 3"BS8F 5BS8F 18BS8H 450BS8H 14BS8H 170BS8F 200BS8F 3"BS8H 14BS8F 1.00"BS8F 1/8"BS8H 6BS8H 6BS8F 400BS8H 1/4"BS8H 20BS8F 270BS8F 45BS8H 12BS8F 7/16"BS8F 1.06"BS8H 40BS8F
57334-606EA 157.34 USD InStock
57334-606 57334-290 57334-294 57334-292 57334-210 57334-298 57334-296 57334-330 57334-610 57334-214 57334-336 57334-212 57334-334 57334-614 57334-339 57334-216 57334-612 57334-280 57334-284 57334-282 57334-200 57334-288 57334-286 57334-324 57334-204 57334-201 57334-202 57334-328 57334-208 57334-326 57334-206 57334-308 57334-309 57334-272 57334-270 57334-276 57334-310 57334-274 57334-314 57334-278 57334-382 57334-262 57334-380 57334-260 57334-266 57334-384 57334-264 57334-302 57334-300 57334-268 57334-306 57334-304 57334-250 57334-372 57334-370 57334-254 57334-376 57334-252 57334-374 57334-258 57334-256 57334-378 57334-360 57334-240 57334-364 57334-244 57334-362 57334-242 57334-368 57334-248 57334-366 57334-246 57334-628 57334-350 57334-199 57334-232 57334-354 57334-230 57334-352 57334-236 57334-632 57334-358 57334-630 57334-234 57334-356 57334-636 57334-634 57334-238 57334-618 57334-616 57334-342 57334-222 57334-340 57334-220 57334-346 57334-226 57334-622 57334-344 57334-224 57334-620 57334-626 57334-348 57334-624 57334-228
VWR® 8" Test Sieves, Brass and Stainless Steel
Sieves Test Sieves
VWR® 8" diameter (203.2 mm ) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a medium sample size is required.

  • Highly durable construction
  • Brass or stainless steel fabrication

Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.

Accessories information: Half height (1") or full height (2") pans are available in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.

Ordering information: Sieves are available in half height (1"), or full height (2"). Sieve height is measured from the frame rim to the sieve cloth. USA Series #12 and higher come standard with UV adhesive, but they may be requested with solder only.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty