Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sieve Receiver Pans, W.S. Tyler™
Supplier: W.S. Tyler
8481 8526 8484 8482 8491 8492 8494
57355-006EA 73.16 USD InStock
57355-006 57355-028 57332-102 57332-100 57355-061 57332-218 57355-040
Sieve Receiver Pans, W.S. Tyler™
Trays/Dishes/Pots Laboratory Trays/Dishes
Sieve Receiver Pans are designed for use with 20.3cm (8") or 30.5cm (12") diameter sieves.
Pans with extended rims may be placed within a stack of sieves when sieving two or more samples simultaneously.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty