Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Bel-Art Quaver® and Quaverette® Vibrating Spatulas, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H36718-0000 H36714-0000 H36715-0000 H36719-0000
13197-376CS 1479.82 USD InStock
13197-376 13197-374 57901-140 57901-106
SP Bel-Art Quaver® and Quaverette® Vibrating Spatulas, Bel-Art Products, a part of SP
Spatulas
Make accurate additions of precious materials as needed and reduce transfer losses.
  • Toothed thumb wheel vibrates tip
  • Flat blade with curved end
  • Quaver® spatula features a flat blade with curved end
Rotating the toothed thumb wheel vibrates the spatula tip, giving you control over transfer from spatula to weighing dish.
The Quaverette® spatula permits utilization of even smaller quantities and containers. Its narrow V-shaped blade and 21 cm (8¼") length maneuvers easily into small mouth containers and tight corners.
Both are available with a plain stainless steel blade or with a brown Fluo-Kem® Teflon® FEP coating.
Certifications: The coating complies with FDA regulations governing direct food contact applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty