Would you like to log in again?
Cuvettes Plus™ Electroporation Cuvettes, BTX™
Supplier: BTX
Cuvettes Plus™
45-0124 450125 450125 45-0124 450126 45-0126
58017-890PK 409.03 USD InStock
CA58017-900 58017-890 CA58017-890 58017-895 CA58017-895 58017-900
Cuvettes Plus™ Electroporation Cuvettes, BTX™
Cuvettes Electroporation Cuvettes
Designed for use in the electroporation of bacteria, yeast, mammalian, and plant cells.
The cuvettes are molded with polished aluminum electrodes and medical grade polycarbonate, washed prior to packaging, and sterilized using gamma irradiation. Gap sizes of 1mm, 2mm, and 4mm are available for high field strengths up to 25.0kV/cm. Each cuvette has a color-coded, leak-resistant round cap that allows single-handed manipulation. Simply discard cuvettes carefully after use. Cuvettes fit all BTX™ and most comparable electroporators, including those from Bio-Rad®, Eppendorf®, and Invitrogen®.
Ordering information: Each sterilized package includes a disposable cuvette and a bonus pipet, which is used for complete removal of the sample after electroporation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty