Would you like to log in again?
CleanTex™ Tex Pad Tapehead Wipes, Advantus
Supplier: CleanTex
CleanTex™
CT801
58019-500CS 631.23 USD InStock
58019-500
CleanTex™ Tex Pad Tapehead Wipes, Advantus
Wipes Disposable Wipes and Towels Presaturated Wipes
Clean dust, dirt, and magnetic oxide buildup from tape heads, guides, capstans, optic fiber connectors and magnetic disc surfaces.
  • Cleans quickly and easily
  • No Residue
  • 100% polyester wipe is non-abrasive
  • Evaporates quickly, so cleaned equipment is ready to use
  • Single-use wipe assures no cross-contamination of equipment
CleanTex™ Tex Pad Tapehead wipes won’t cause any abrasion on discs and are also lint-free, leaving no residue.The pre-saturated 91% isopropyl alcohol and 9% deionized water solution evaporates quickly without a trace, leaving equipment ready to use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty