Would you like to log in again?
AlcoPad® Electronic Component Cleaning Pad, CleanTex™
Supplier: CleanTex
AlcoPad®
CT806
58019-507CS 639.99 USD InStock
58019-507
AlcoPad® Electronic Component Cleaning Pad, CleanTex™
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Disposable pad for precision industrial cleaning and removal of magnetic oxides. Presaturated with 91% reagent-grade isopropyl alcohol and 9% deionized water.
  • Ultrafiltered to 0.2 µ
  • Non corrosive, non abrasive and lint-free
  • Each pad is hermetically sealed and individually wrapped
Recommended for cleaning machine parts, electronic components, magnetic tape heads, electro-optics, PC boards, bearings and test and lab equipment.
Packaging: Packed in boxes of 80 wipers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty