Would you like to log in again?
OpticPad® Optical Surface Cleaning Pads, CleanTex™
Supplier: CleanTex
OpticPad®
CT811
58019-512CS 738.62 USD InStock
58019-512
OpticPad® Optical Surface Cleaning Pads, CleanTex™
Wipes Disposable Wipes and Towels Lens Papers
Disposable, lint-free pad is presaturated for cleaning optical-grade surfaces.
Ideal for cleaning and polishing front and back surfaces of mirrors, microscope lenses, safety goggles, and photographic equipment. Solvent is filtered to 0.2 µm and evaporates quickly without leaving a residue.
Packaging: Each pad is hermetically sealed and individually wrapped.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty