Would you like to log in again?
Wet & Dry™ Presaturated Pads, CleanTex™
Supplier: CleanTex
Wet & Dry™
CT812
58019-514CS 672.08 USD InStock
58019-514
Wet & Dry™ Presaturated Pads, CleanTex™
Wipes Disposable Wipes and Towels Presaturated Wipes
Use this system for cleaning, drying and controlling static on terminal, CRT and COM screens, display screens and instrument glass.
Not for use on nylon mesh antiglare screens.
Two pads are hermetically sealed in separate foil packets and attached in the middle. The wet pad is moistened with a special antistatic screen cleaning solution (ultrafiltered to 0.2 µl) that neutralizes electrostatic charges while removing contaminants from the screen. Made from 100% lint-free polyester spunbonded fabric that contains no binders or additives to affect surface being cleaned. The dry pad is a pure, nonwoven cloth combining the absorbency of cellulose with the strength of polyester. This high absorbency drying pad polishes screens to a streak-free finish.
Packaging: There are 80 pads (40 twinpaks) per package.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty