Would you like to log in again?
Stainless Steel Cleanroom Tables, Perforated Top, Eagle MHC™
Supplier: Eagle MHC
CRPT2448T CRPT2436T-C CRPT3660T-C CRPT3696T-C CRPT3684T CRPT3072T CRPT3036T-C CRPT2484T CRPT3060T CRPT3672T-C CRPT2460T CRPT2472T CRPT2496T
58202-408EA 8589.09 USD InStock
58202-408 58202-750 58202-754 58202-402 58202-952 58202-744 58202-400 58202-752 58202-406 58202-748 58202-404 58202-954 58202-746
Stainless Steel Cleanroom Tables, Perforated Top, Eagle MHC™
Furniture Tables and Workbenches
14-gauge, Type 304 stainless steel top.
  • Perforated top allows for 40% laminar flow
  • Knocked down stainless steel C-frame
Brushed stainless top features ¾" holes on 1" centers.
Ordering information: Alcohol wiped and cleanroom bagged. Shipped knocked top reducing freight costs and potential for freight damage. Other base frame options available; contact your Avantor sales representative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty