Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Wall-Mounted Gowning Rack, Eagle MHC
Supplier: Eagle MHC
EP2448-WGR EP2472-WGR C2460-WGR C2448-WGR C2472-WGR EP2460-WGR
58202-130EA 1193.59 USD InStock
58202-130 58202-662 58202-596 58202-652 58202-642 58202-602
Wall-Mounted Gowning Rack, Eagle MHC
Furniture Gowning Racks
Wall mounted gowning rack incorporates wire shelving components that offer a more economical rack option for less demanding environments.
  • Open wire construction facilitates laminar flow
  • Convenient for storage of lab coats, uniforms and garments
Features hanger slots on 2½" centers and 24" wide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty